- Purification of Maize Nucleotide Pyrophosphatase/Phosphodiesterase Casts Doubt on the Existence of Zeatin
Purification of Maize Nucleotide Pyrophosphatase/Phosphodiesterase Casts Doubt on the Existence of Zeatin
Frontiers in plant science (2017-09-08)
Tomáš Hluska, Marek Šebela, René Lenobel, Ivo Frébort, Petr Galuszka
PMID28878803
ABSTRACT
Almost 25 years ago, an enzyme named zeatin