Merck
m
Nature (2017-08-10)
Hua-Bing Li, Jiyu Tong, Shu Zhu, Pedro J Batista, Erin E Duffy, Jun Zhao, Will Bailis, Guangchao Cao, Lina Kroehling, Yuanyuan Chen, Geng Wang, James P Broughton, Y Grace Chen, Yuval Kluger, Matthew D Simon, Howard Y Chang, Zhinan Yin, Richard A Flavell
ABSTRACT
N
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Socs1
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ~98% (HPLC)