Merck
Monoanionic
Nuclear medicine and biology (2017-01-23)
Malgorzata Lipowska, Jeffrey Klenc, Nashwa Jarkas, Luigi G Marzilli, Andrew T Taylor
ABSTRACT
New tracers lacking the negatively charged pendant carboxyl group previously thought to be essential for rapid renal extraction,
Sigma-Aldrich
Dimethyl iminodiacetate hydrochloride, ≥98.0%