Merck
  • Glycoprotein 2 is a specific cell surface marker of human pancreatic progenitors.
Glycoprotein 2 is a specific cell surface marker of human pancreatic progenitors.
Nature communications (2017-08-25)
Kathryn F Cogger, Ankit Sinha, Farida Sarangi, Emily C McGaugh, Diane Saunders, Craig Dorrell, Salvador Mejia-Guerrero, Yasaman Aghazadeh, Jillian L Rourke, Robert A Screaton, Markus Grompe, Philip R Streeter, Alvin C Powers, Marcela Brissova, Thomas Kislinger, M Cristina Nostro
ABSTRACT
PDX1
Sigma-Aldrich
SANT-1, ≥98% (HPLC), powder
Sigma-Aldrich
Pyrvinium pamoate salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Triton X-100, laboratory grade