Merck
Fludarabine Inhibits K
Frontiers in pharmacology (2017-04-15)
Alicia de la Cruz, Alba Vera-Zambrano, Diego A Peraza, Carmen Valenzuela, Juan M Zapata, Gema Perez-Chacon, Teresa Gonzalez
ABSTRACT
Fludarabine (F-ara-A) is a purine analog commonly used in the treatment of indolent B cell malignancies that interferes with different aspects of DNA and RNA synthesis. K
Sigma-Aldrich
5-(4-Phenoxybutoxy)psoralen, ≥98% (HPLC), solid
Sigma-Aldrich
2-Fluoroadenine-9-β-D-arabinofuranoside, DNA synthesis and methylation inhibitor