Merck
  • Why choose tiotropium for my patient? A comprehensive review of actions and outcomes versus other bronchodilators.
Why choose tiotropium for my patient? A comprehensive review of actions and outcomes versus other bronchodilators.
Respiratory medicine (2017-06-15)
David M Halpin, Alan G Kaplan, Richard K Russell
ABSTRACT
Chronic obstructive pulmonary disease (COPD) and asthma are leading causes of morbidity and mortality. This narrative review provides an appraisal of the pharmacological and clinical characteristics of tiotropium in COPD and asthma, and examines how these compare with other long-acting bronchodilators. The evidence base is placed into context by relating it to factors affecting clinicians' choice of therapy. Desirable attributes of a long-acting muscarinic antagonist (LAMA) maintenance therapy include effective pharmacological bronchodilation, improved lung function, exacerbation efficacy, and positive effects on symptom control, exercise capacity and quality of life across a broad patient population. Tolerability and convenience of use are also important for patient well-being and treatment adherence. Tiotropium shows higher affinity for muscarinic receptors than ipratropium, and prolonged binding to the M With over 10 years' prescribing history and 50 million patient-years of use, tiotropium has the benefit of a more extensive clinical evidence base than other long-acting bronchodilators, with demonstrated efficacy and safety in COPD and symptomatic asthma.
Sigma-Aldrich
5,5-Dimethylhydantoin, 97%