Merck
  • Comparison of derivatization/ionization techniques for liquid chromatography tandem mass spectrometry analysis of oxylipins.
Comparison of derivatization/ionization techniques for liquid chromatography tandem mass spectrometry analysis of oxylipins.
Prostaglandins & other lipid mediators (2017-02-22)
Sven W Meckelmann, Stefan Hellhake, Maryvonne Steuck, Michael Krohn, Nils Helge Schebb
ABSTRACT
The performance of two derivatization and ionization techniques for the quantitative reversed phase liquid chromatography (LC)- mass spectrometry (MS) analysis of hydroxy fatty acids (OH-PUFA) in plasma was evaluated: One used AMPP (N-(4-aminomethylphenyl)pyridinium chloride) leading to a positive charged amid-derivate which can be detected by electrospray ionization (ESI)-MS. Second yielded penta fluorobenzyl bromide (PFB) ester derivates allowing detection in electron capture atmospheric pressure chemical ionization (ecAPCI)-MS. The sensitivity of detection of a comprehensive set of hydroxy fatty acids of n6- and n3- poly unsaturated fatty acids was investigated. On the SCIEX3200 MS the applied PFB derivatization led to poor limits of detection (LOD) of 10-100nM (0.1-1pmol/0.03-0.3ng on column). By contrast, AMPP derivatization led to a similar sensitivity compared to the standard ESI(-) of non derivatized analytes (LOD about 1nM (10fmol/3pg on column)). For several analytes, including 9-HETE, 11-HETE and 17-HDHA the AMPP derivatization improved sensitivity enabling their detection in human plasma. However, precision was reduced by AMPP derivatization and variation in IS recovery indicated a strong matrix influence on the MS-signal. In sum, with the instrumentation used, neither of these derivatization methods improves in our hands the LC-MS based quantification of oxylipins.
Sigma-Aldrich
4-[(N-Boc)aminomethyl]aniline, 97%
Sigma-Aldrich
Ethanol, puriss. p.a., absolute, ≥99.8% (GC)