Merck
  • An isotope-dilution UPLC-MS/MS technique for the human biomonitoring of the internal exposure to glycidol via a valine adduct at the N-terminus of hemoglobin.
An isotope-dilution UPLC-MS/MS technique for the human biomonitoring of the internal exposure to glycidol via a valine adduct at the N-terminus of hemoglobin.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2017-05-30)
Jan Hielscher, Bernhard H Monien, Klaus Abraham, Sönke Jessel, Albrecht Seidel, Alfonso Lampen
ABSTRACT
Fatty acid esters of glycidol (glycidyl esters) are processing contaminants generated as a byproduct of the industrial deodorization of vegetable oils and fats. Oral intake of glycidyl esters leads to the release of glycidol in the gastrointestinal tract. Glycidol is carcinogenic, genotoxic and teratogenic in rodents. It is rated as probably carcinogenic to humans (IARC group 2A). The determination of internal exposure of glycidol may support the assessment of the possible human health risks related to glycidyl ester intake. For this purpose, hemoglobin adducts of glycidol may be suitable biomarkers reflecting the cumulative exposure of up to four months. We applied a modified Edman degradation to assess the glycidol adduct at the N-terminal valine, N-(2,3-dihydroxypropyl)-valine (2,3-diHOPr-Val), of hemoglobin. The modified valine was cleaved with fluorescein-5-isothiocyanate (FITC), resulting in the formation of N-(2,3-dihydroxypropyl)-valine fluorescein thiohydantoin (DHP-Val-FTH). An isotope-dilution technique was developed for the quantification of the thiohydantoin analyte by ultra performance liquid chromatography-tandem mass spectrometry (UPLC-MS/MS) and DHP-Val-d
Sigma-Aldrich
Glycidol, 96%
Pricing and availability is not currently available.
Sigma-Aldrich
L-Valine-d8, ≥98 atom % D, ≥98% (CP)
Pricing and availability is not currently available.