Merck
  • Facile preparation of water-soluble hyperbranched polyamine functionalized multiwalled carbon nanotubes for high-efficiency organic dye removal from aqueous solution.
Facile preparation of water-soluble hyperbranched polyamine functionalized multiwalled carbon nanotubes for high-efficiency organic dye removal from aqueous solution.
Scientific reports (2017-06-18)
Lihua Hu, Zhongping Yang, Yaoguang Wang, Yan Li, Dawei Fan, Di Wu, Qin Wei, Bin Du
ABSTRACT
Water-soluble hyperbranched polyamine functionalized multiwalled carbon nanotubes nanocomposite (WHPA-OMCNT) was successfully prepared and applied to water remediation in this paper. WHPA-OMCNT was characterized by Fourier transform infrared spectroscopy (FTIR), thermogravimetric analysis (TGA), zeta potential, scanning electron microscopy (SEM) and transmission electron microscope (TEM) analyses. WHPA-OMCNT exhibited excellent adsorption performance for removal of organic dyes e.g., methylene blue (MB), malachite green (MG) and methyl violet (MV). The equilibrium adsorption capacity was 800.0 mg g
Sigma-Aldrich
N-Ethylethylenediamine, 98%