Merck
  • Enabling Prussian Blue with Tunable Localized Surface Plasmon Resonances: Simultaneously Enhanced Dual-Mode Imaging and Tumor Photothermal Therapy.
Enabling Prussian Blue with Tunable Localized Surface Plasmon Resonances: Simultaneously Enhanced Dual-Mode Imaging and Tumor Photothermal Therapy.
ACS nano (2016-12-28)
Xiaojun Cai, Wei Gao, Linlin Zhang, Ming Ma, Tianzhi Liu, Wenxian Du, Yuanyi Zheng, Hangrong Chen, Jianlin Shi
ABSTRACT
Prussian blue (PB) has been used as a photothermal conversion agent to generate heat to induce localized damage to tumor. However, its therapeutic efficiency is far from satisfactory. One of the major obstacles is that the maximum NIR absorption peak of PB within 690-720 nm cannot be optimized near the wavelength of the laser to enhance its therapeutic efficiency. Herein, we report that the integration of Gd