Merck
  • Occurrence, distribution and risk assessment of suspected endocrine-disrupting chemicals in surface water and suspended particulate matter of Yangtze River (Nanjing section).
Occurrence, distribution and risk assessment of suspected endocrine-disrupting chemicals in surface water and suspended particulate matter of Yangtze River (Nanjing section).
Ecotoxicology and environmental safety (2016-10-11)
Yan-Hua Liu, Sheng-Hu Zhang, Gui-Xiang Ji, Sheng-Min Wu, Rui-Xin Guo, Jie Cheng, Zheng-Yu Yan, Jian-Qiu Chen
ABSTRACT
The occurrence and distribution of eight selected endocrine-disrupting chemicals were investigated in samples of surface water and suspended particulate matter (SPM) in Nanjing section of Yangtze River over a year (the flow period, the wet period and the dry period). All target compounds were detected at least once in surface water with 4-tert-butylphenol (4-TBP), nonyphenol (NP) and bisphenol A (BPA) as the dominant compounds, with concentrations in the range of 225-1121ng/L, 1.4-858ng/L and 1.7-563ng/L, respectively. Except for December, all selected compounds for the other sampling times were not found in all sampling points. NP (mean concentration 69.8µg/g) and BPA (mean concentration 51.8µg/g) were also the dominant estrogens in SPM. In addition, the highest total compounds concentrations were found in December in both phases, which could be due to the low flow conditions and temperature during this season. Meanwhile, a significant positive correlation was found between the total compounds concentrations in the water phase and those in SPM phase. Risk assessment based on the calculated risk quotients (RQ) showed that low and moderate risk for the aquatic environment from presence of the target compounds at all sampling points with exception of 4-TBP and NP which might pose a high risk to aquatic organisms.
Sigma-Aldrich
4-Butylphenol, AldrichCPR