Merck
  • Mfge8 regulates enterocyte lipid storage by promoting enterocyte triglyceride hydrolase activity.
Mfge8 regulates enterocyte lipid storage by promoting enterocyte triglyceride hydrolase activity.
JCI insight (2016-11-05)
Amin Khalifeh-Soltani, Deepti Gupta, Arnold Ha, Jahangir Iqbal, Mahmood Hussain, Michael J Podolsky, Kamran Atabai
ABSTRACT
The small intestine has an underappreciated role as a lipid storage organ. Under conditions of high dietary fat intake, enterocytes can minimize the extent of postprandial lipemia by storing newly absorbed dietary fat in cytoplasmic lipid droplets. Lipid droplets can be subsequently mobilized for the production of chylomicrons. The mechanisms that regulate this process are poorly understood. We report here that the milk protein Mfge8 regulates hydrolysis of cytoplasmic lipid droplets in enterocytes after interacting with the αvβ3 and αvβ5 integrins. Mice deficient in Mfge8 or the αvβ3 and αvβ5 integrins accumulate excess cytoplasmic lipid droplets after a fat challenge. Mechanistically, interruption of the Mfge8-integrin axis leads to impaired enterocyte intracellular triglyceride hydrolase activity in vitro and in vivo. Furthermore, Mfge8 increases triglyceride hydrolase activity through a PI3 kinase/mTORC2-dependent signaling pathway. These data identify a key role for Mfge8 and the αvβ3 and αvβ5 integrins in regulating enterocyte lipid processing.
Sigma-Aldrich
Anti-Perilipin-3, C-terminus Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid