Merck
  • Membrane-sensitive conformational states of helix 8 in the metabotropic Glu2 receptor, a class C GPCR.
Membrane-sensitive conformational states of helix 8 in the metabotropic Glu2 receptor, a class C GPCR.
PloS one (2012-08-08)
Agostino Bruno, Gabriele Costantino, Gianni de Fabritiis, Manuel Pastor, Jana Selent
ABSTRACT
The recent elucidation of the X-ray structure of several class A GPCRs clearly indicates that the amphipathic helix 8 (H8) is a conserved structural domain in most crystallized GPCRs. Very little is known about the presence and the possible role of an analogous H8 domain in the distantly related class C GPCRs. In this study, we investigated the structural properties for the H8 domain of the mGluR2 receptor, a class C GPCR, by applying extended molecular dynamics simulations. Our study indicates that the amphipathic H8 adopts membrane-sensitive conformational states, which depend on the membrane composition. Cholesterol-rich membranes stabilize the helical structure of H8 whereas cholesterol-depleted membranes induce a disruption of H8. The observed link between membrane cholesterol levels and H8 conformational states suggests that H8 behaves as a sensor of cholesterol concentration.