Merck
  • Mast cell degranulation is negatively regulated by the Munc13-4-binding small-guanosine triphosphatase Rab37.
Mast cell degranulation is negatively regulated by the Munc13-4-binding small-guanosine triphosphatase Rab37.
Scientific reports (2016-03-05)
Hironori Higashio, Yoh-ichi Satoh, Tomoyuki Saino
ABSTRACT
Mast cell degranulation is regulated by the small guanosine triphosphatases (GTPases) Rab27a and Rab27b, which have distinct and opposing roles: Rab27b acts as a positive regulator through its effector protein Munc13-4, a non-neuronal isoform of the vesicle-priming Munc13 family of proteins, whereas Rab27a acts as a negative regulator through its effector protein melanophilin, by maintaining integrity of cortical filamentous actin (F-actin), a barrier to degranulation. Here we investigated the role of Rab37, one of the Rab GTPases assumed to be implicated in regulated secretion during mast cell degranulation. Using the RBL-2H3 mast cell line, we detected Rab37 on the secretory granules and found that antigen-induced degranulation was extensively increased by either knockdown of Rab37 or overexpression of a dominant-active Rab37 mutant. This hypersecretion phenotype in the Rab37-knockdown cells was suppressed by simultaneous knockdown of Rab27a and Rab27b or of Munc13-4, but not by disruption of cortical F-actin. We further found that Rab37 interacted with Munc13-4 in a GTP-independent manner and formed a Rab27-Munc13-4-Rab37 complex. These results suggest that Rab37 is a Munc13-4-binding protein that inhibits mast cell degranulation through its effector protein, by counteracting the vesicle-priming activity of the Rab27-Munc13-4 system.
Sigma-Aldrich
Anti-Rab27 (N-terminal) antibody produced in rabbit, ~1.5 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution