Merck
  • Cobalt iron oxide nanoparticles induce cytotoxicity and regulate the apoptotic genes through ROS in human liver cells (HepG2).
Cobalt iron oxide nanoparticles induce cytotoxicity and regulate the apoptotic genes through ROS in human liver cells (HepG2).
Colloids and surfaces. B, Biointerfaces (2016-11-05)
Maqusood Ahamed, Mohd Javed Akhtar, M A Majeed Khan, Hisham A Alhadlaq, Aws Alshamsan
ABSTRACT
Cobalt iron oxide (CoFe
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Sigma-Aldrich
Cobalt iron oxide, nanopowder, 30 nm particle size (TEM), 99% trace metals basis