Merck
  • Suppression of STAT3 and HIF-1 alpha mediates anti-angiogenic activity of betulinic acid in hypoxic PC-3 prostate cancer cells.
Suppression of STAT3 and HIF-1 alpha mediates anti-angiogenic activity of betulinic acid in hypoxic PC-3 prostate cancer cells.
PloS one (2011-07-07)
Jimin Shin, Hyo-Jeong Lee, Deok-Beom Jung, Ji Hoon Jung, Hyo-Jung Lee, Eun-Ok Lee, Seok Geun Lee, Beom Sang Shim, Seung Hoon Choi, Seong Gyu Ko, Kwang Seok Ahn, Soo-Jin Jeong, Sung-Hoon Kim
ABSTRACT
Signal transducer and activator of transcription 3 (STAT3) is a transcription factor that regulates various cellular processes such as cell survival, angiogenesis and proliferation. In the present study, we examined that betulinic acid (BA), a triterpene from the bark of white birch, had the inhibitory effects on hypoxia-mediated activation of STAT3 in androgen independent human prostate cancer PC-3 cells. BA inhibited the protein expression and the transcriptional activities of hypoxia-inducible factor-1α (HIF-1α) under hypoxic condition. Consistently, BA blocked hypoxia-induced phosphorylation, DNA binding activity and nuclear accumulation of STAT3. In addition, BA significantly reduced cellular and secreted levels of vascular endothelial growth factor (VEGF), a critical angiogenic factor and a target gene of STAT3 induced under hypoxia. Furthermore, BA prevented in vitro capillary tube formation in human umbilical vein endothelial cells (HUVECs) maintained in conditioned medium of hypoxic PC-3 cells, implying anti-angiogenic activity of BA under hypoxic condition. Of note, chromatin immunoprecipitation (ChiP) assay revealed that BA inhibited binding of HIF-1α and STAT3 to VEGF promoter. Furthermore, silencing STAT3 using siRNA transfection effectively enhanced the reduced VEGF production induced by BA treatment under hypoxia. Taken together, our results suggest that BA has anti-angiogenic activity by disturbing the binding of HIF-1α and STAT3 to the VEGF promoter in hypoxic PC-3 cells.
Sigma-Aldrich
Betulinic acid, ≥98% (HPLC)
Sigma-Aldrich
EZ-Zyme Chromatin Prep Kit, Contains proprietary reagents optimized for the enzymatic shearing of chromatin from mammalian cells at higher resolution than sonication for use in chromatin immunoprecipitation (ChIP) assays.