Merck
  • Stereoselective [4 + 1] annulation reactions with silyl vinylketenes derived from Fischer carbene complexes.
Stereoselective [4 + 1] annulation reactions with silyl vinylketenes derived from Fischer carbene complexes.
The Journal of organic chemistry (2006-08-12)
William H Moser, Laura A Feltes, Liangdong Sun, Matthew W Giese, Ryan W Farrell
ABSTRACT
Stable silyl vinylketenes were prepared via the thermal reaction of Fischer carbene complexes with triisopropylsilyl- or tert-butyldimethylsilyl-substituted alkynes. The ability of these silyl vinylketenes to participate with carbenoid reagents in [4 + 1] annulation reactions was investigated. The best results were obtained with diazomethane and substituted diazomethane reagents, which provided cyclopentenone products in excellent yields and essentially complete stereoselectivity.
Sigma-Aldrich
Triisopropylsilyl chloride, 97%