Merck
  • Disruption of murine mp29/Syf2/Ntc31 gene results in embryonic lethality with aberrant checkpoint response.
Disruption of murine mp29/Syf2/Ntc31 gene results in embryonic lethality with aberrant checkpoint response.
PloS one (2012-03-27)
Chia-Hsin Chen, Po-Chen Chu, Liekyeow Lee, Huang-Wei Lien, Tse-Ling Lin, Chi-Chen Fan, Peter Chi, Chang-Jen Huang, Mau-Sun Chang
ABSTRACT
Human p29 is a putative component of spliceosomes, but its role in pre-mRNA is elusive. By siRNA knockdown and stable overexpression, we demonstrated that human p29 is involved in DNA damage response and Fanconi anemia pathway in cultured cells. In this study, we generated p29 knockout mice (mp29(GT/GT)) using the mp29 gene trap embryonic stem cells to study the role of mp29 in DNA damage response in vivo. Interruption of mp29 at both alleles resulted in embryonic lethality. Embryonic abnormality occurred as early as E6.5 in mp29(GT/GT) mice accompanied with decreased mRNA levels of α-tubulin and Chk1. The reduction of α-tubulin and Chk1 mRNAs is likely due to an impaired post-transcriptional event. An aberrant G2/M checkpoint was found in mp29 gene trap embryos when exposed to aphidicolin and UV light. This embryonic lethality was rescued by crossing with mp29 transgenic mice. Additionally, the knockdown of zfp29 in zebrafish resulted in embryonic death at 72 hours of development postfertilization (hpf). A lower level of acetylated α-tubulin was also observed in zfp29 morphants. Together, these results illustrate an indispensable role of mp29 in DNA checkpoint response during embryonic development.
Sigma-Aldrich
MISSION® esiRNA, targeting human SYF2