Merck
  • Plasmacytoid dendritic cells capture and cross-present viral antigens from influenza-virus exposed cells.
Plasmacytoid dendritic cells capture and cross-present viral antigens from influenza-virus exposed cells.
PloS one (2009-09-23)
Gabrielle Lui, Olivier Manches, Juliette Angel, Jean-Paul Molens, Laurence Chaperot, Joël Plumas
ABSTRACT
Among the different subsets of dendritic cells (DC), plasmacytoid dendritic cells (PDC) play a unique role in secreting large amounts of type I interferons upon viral stimulation, but their efficiency as antigen-presenting cells has not been completely characterized. We show here, by flow cytometry, with human primary blood PDC and with a PDC cell line, that PDC display poor endocytic capacity for soluble or cellular antigens when compared to monocyte-derived myeloid DC. However, immature PDC efficiently take up cellular material from live influenza-exposed cells, subsequently mature and cross-present viral antigens very efficiently to specific CD8+ T cells. Therefore, during viral infection PDC not only secrete immunomodulatory cytokines, but also recognize infected cells and function as antigen cross-presenting cells to trigger the anti-viral immune response.
Sigma-Aldrich
Anti-Integrin αVβ3 Antibody, clone LM609, clone LM609, Chemicon®, from mouse
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Integrin αVβ5 Antibody, clone P1F6, clone P1F6, Chemicon®, from mouse
Pricing and availability is not currently available.