Merck
  • A Hereditary Enteropathy Caused by Mutations in the SLCO2A1 Gene, Encoding a Prostaglandin Transporter.
A Hereditary Enteropathy Caused by Mutations in the SLCO2A1 Gene, Encoding a Prostaglandin Transporter.
PLoS genetics (2015-11-06)
Junji Umeno, Tadakazu Hisamatsu, Motohiro Esaki, Atsushi Hirano, Naoya Kubokura, Kouichi Asano, Shuji Kochi, Shunichi Yanai, Yuta Fuyuno, Katsuyoshi Shimamura, Naoki Hosoe, Haruhiko Ogata, Takashi Watanabe, Kunihiko Aoyagi, Hidehisa Ooi, Kenji Watanabe, Shigeyoshi Yasukawa, Fumihito Hirai, Toshiyuki Matsui, Mitsuo Iida, Tsuneyoshi Yao, Toshifumi Hibi, Kenjiro Kosaki, Takanori Kanai, Takanari Kitazono, Takayuki Matsumoto
ABSTRACT
Previously, we proposed a rare autosomal recessive inherited enteropathy characterized by persistent blood and protein loss from the small intestine as chronic nonspecific multiple ulcers of the small intestine (CNSU). By whole-exome sequencing in five Japanese patients with CNSU and one unaffected individual, we found four candidate mutations in the SLCO2A1 gene, encoding a prostaglandin transporter. The pathogenicity of the mutations was supported by segregation analysis and genotyping data in controls. By Sanger sequencing of the coding regions, 11 of 12 other CNSU patients and 2 of 603 patients with a diagnosis of Crohn's disease were found to have homozygous or compound heterozygous SLCO2A1 mutations. In total, we identified recessive SLCO2A1 mutations located at seven sites. Using RT-PCR, we demonstrated that the identified splice-site mutations altered the RNA splicing, and introduced a premature stop codon. Tracer prostaglandin E2 uptake analysis showed that the mutant SLCO2A1 protein for each mutation exhibited impaired prostaglandin transport. Immunohistochemistry and immunofluorescence analyses revealed that SLCO2A1 protein was expressed on the cellular membrane of vascular endothelial cells in the small intestinal mucosa in control subjects, but was not detected in affected individuals. These findings indicate that loss-of-function mutations in the SLCO2A1 gene encoding a prostaglandin transporter cause the hereditary enteropathy CNSU. We suggest a more appropriate nomenclature of "chronic enteropathy associated with SLCO2A1 gene" (CEAS).
Sigma-Aldrich
Anti-SLCO2A1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution