Merck
  • Stabilized Conversion Efficiency and Dye-Sensitized Solar Cells from Beta vulgaris Pigment.
Stabilized Conversion Efficiency and Dye-Sensitized Solar Cells from Beta vulgaris Pigment.
International journal of molecular sciences (2013-02-23)
Angel Ramon Hernández-Martínez, Miriam Estévez, Susana Vargas, Rogelio Rodríguez
ABSTRACT
Dye-Sensitized Solar Cells (DSSCs), based on TiO(2) and assembled using a dye from Beta vulgaris extract (BVE) with Tetraethylorthosilicate (TEOS), are reported. The dye BVE/TEOS increased its UV resistance, rendering an increase in the cell lifetime; the performance of these solar cells was compared to those prepared with BVE without TEOS. The efficiency η for the solar energy conversion was, for BVE and BVE/TEOS, of 0.89% ± 0.006% and 0.68% ± 0.006% with a current density Jsc of 2.71 ± 0.003 mA/cm2 and 2.08 ± 0.003 mA/cm2, respectively, using in both cases an irradiation of 100 mW/cm2 at 25 °C. The efficiency of the BVE solar cell dropped from 0.9 ± 0.006 to 0.85 ± 0.006 after 72 h of operation, whereas for the BVE/TEOS, the efficiency remained practically constant in the same period of time.
Sigma-Aldrich
3-Methoxypropionitrile, ≥98.0% (GC)