Merck
  • The intramembrane protease SPPL2a promotes B cell development and controls endosomal traffic by cleavage of the invariant chain.
The intramembrane protease SPPL2a promotes B cell development and controls endosomal traffic by cleavage of the invariant chain.
The Journal of experimental medicine (2012-12-26)
Janna Schneppenheim, Ralf Dressel, Susann Hüttl, Renate Lüllmann-Rauch, Michael Engelke, Kai Dittmann, Jürgen Wienands, Eeva-Liisa Eskelinen, Irm Hermans-Borgmeyer, Regina Fluhrer, Paul Saftig, Bernd Schröder
ABSTRACT
Regulated intramembrane proteolysis is a central cellular process involved in signal transduction and membrane protein turnover. The presenilin homologue signal-peptide-peptidase-like 2a (SPPL2a) has been implicated in the cleavage of type 2 transmembrane proteins. We show that the invariant chain (li, CD74) of the major histocompatability class II complex (MHCII) undergoes intramembrane proteolysis mediated by SPPL2a. B lymphocytes of SPPL2a(-/-) mice accumulate an N-terminal fragment (NTF) of CD74, which severely impairs membrane traffic within the endocytic system and leads to an altered response to B cell receptor stimulation, reduced BAFF-R surface expression, and accumulation of MHCII in transitional developmental stage T1 B cells. This results in significant loss of B cell subsets beyond the T1 stage and disrupted humoral immune responses, which can be recovered by additional ablation of CD74. Hence, we provide evidence that regulation of CD74-NTF levels by SPPL2a is indispensable for B cell development and function by maintaining trafficking and integrity of MHCII-containing endosomes, highlighting SPPL2a as a promising pharmacological target for depleting and/or modulating B cells.
Sigma-Aldrich
Anti-CD3, T Cell antibody produced in rabbit, whole antiserum
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
500 mg
Estimated to ship on 23 March 2025
US$392.00
To order products, please contact your local dealer.