MilliporeSigma
  • The role of insulin in the response of murine T lymphocytes to mitogenic stimulation in vitro.
The role of insulin in the response of murine T lymphocytes to mitogenic stimulation in vitro.
Journal of immunology (Baltimore, Md. : 1950) (1980-02-01)
E C Snow, T L Feldbush, J A Oaks
ABSTRACT
The ability of insulin to influence the responsiveness of murine T lymphocytes in a culture system containing a serum substitute was documented. The presence of insulin was found to enhance the concanavalin A (Con A) reactivity of the lymphocytes. Once the cells were activated by a short-term exposure to Con A, insulin was capable of replacing Con A for the continued stimulation of the cells. This was true both for lymphocyte proliferation and for the generation of nonspecific cytotoxic T lymphocytes. The presence or absence of insulin was not found to influence the phytohemagglutinin responsiveness of the T lymphocytes. Possible reasons for the observed results are discussed in relation to a proposed model for lymphocyte activation.