Merck
  • Abscisic-acid-induced cellular apoptosis and differentiation in glioma via the retinoid acid signaling pathway.
Abscisic-acid-induced cellular apoptosis and differentiation in glioma via the retinoid acid signaling pathway.
International journal of cancer (2015-11-26)
Nan Zhou, Yu Yao, Hongxing Ye, Wei Zhu, Liang Chen, Ying Mao
ABSTRACT
Retinoid acid (RA) plays critical roles in regulating differentiation and apoptosis in a variety of cancer cells. Abscisic acid (ABA) and RA are direct derivatives of carotenoids and share structural similarities. Here we proposed that ABA may also play a role in cellular differentiation and apoptosis by sharing a similar signaling pathway with RA that may be involved in glioma pathogenesis. We reported for the first time that the ABA levels were twofold higher in low-grade gliomas compared with high-grade gliomas. In glioma tissues, there was a positive correlation between the ABA levels and the transcription of cellular retinoic acid-binding protein 2 (CRABP2) and a negative correlation between the ABA levels and transcription of fatty acid-binding protein 5 (FABP5). ABA treatment induced a significant increase in the expression of CRABP2 and a decrease in the expression of peroxisome proliferator-activated receptor (PPAR) in glioblastoma cells. Remarkably, both cellular apoptosis and differentiation were increased in the glioblastoma cells after ABA treatment. ABA-induced cellular apoptosis and differentiation were significantly reduced by selectively silencing RAR-α, while RAR-α overexpression exaggerated the ABA-induced effects. These results suggest that ABA may play a role in the pathogenesis of glioma by promoting cellular apoptosis and differentiation through the RA signaling pathway.
Sigma-Aldrich
γ-Globulins from bovine blood, ≥99% (agarose gel electrophoresis)
Sigma-Aldrich
(+)-Abscisic acid, ≥98% (HPLC)