Merck
  • TOXOPLASMA AND VIRAL ANTIBODIES AMONG HIV PATIENTS AND INMATES IN CENTRAL JAVA, INDONESIA.
TOXOPLASMA AND VIRAL ANTIBODIES AMONG HIV PATIENTS AND INMATES IN CENTRAL JAVA, INDONESIA.
The Southeast Asian journal of tropical medicine and public health (2016-02-13)
Yulia Sari, Sri Haryati, Irvan Raharjo, Afiono Agung Prasetyo
ABSTRACT
In Indonesia, Toxoplasma and its associations with blood-borne viruses have been poorly studied. In order to study the association between anti-Toxoplasma antibodies and blood-borne viral antibodies, blood samples from 497 participants (375 inmates from four prisons in Central Java, Indonesia and 122 HIV patients at a Voluntary Counseling and Testing Clinic in Surakarta, Indonesia) were tested for serological markers of Toxoplasma, human immunodeficiency virus (HIV), hepatitis B virus (HBV), hepatitis C virus (HCV), hepatitis D virus (HDV) and human T-lymphotropic virus types I and II (HTLV-1/2). Anti-Toxoplasma IgG and IgM positivity rates were 41.6% and 3.6%, respectively. One point two percent of participants was positive for both anti-Toxoplasma IgG and IgM antibodies. Sixteen point five percent, 11.3%, 2.6% and 2.8% of participants were positive for anti- Toxoplasma IgG combined with anti-HCV antibodies, anti-Toxoplasma IgG combined with anti-HIV antibodies, anti-Toxoplasma IgM combined with anti-HIV antibodes and anti-Toxoplasma IgG combined with both anti-HIV and anti-HCV antibodies, respectively. Anti-Toxoplasma IgM seropositivity was associated with anti-HIV (aOR = 4.3; 95% CI: 1.112-16.204, p = 0.034). Anti-Toxoplasma IgG antibodies were associated with anti-HCV (aOR = 2.8; 95% CI: 1.749-4.538, p < 0.001) and history of injection drug use (aOR = 3.1; 95% CI: 1.905-5.093, p < 0.001). In conclusion, we recommend patients with HIV, HCV infection and injection drug users should be screened for Toxoplasma infection in Indonesia.