Merck
  • Gold film-catalysed benzannulation by microwave-assisted, continuous flow organic synthesis (MACOS).
Gold film-catalysed benzannulation by microwave-assisted, continuous flow organic synthesis (MACOS).
Beilstein journal of organic chemistry (2009-09-25)
Gjergji Shore, Michael Tsimerman, Michael G Organ
ABSTRACT
Methodology has been developed for laying down a thin gold-on-silver film on the inner surface of glass capillaries for the purpose of catalysing benzannulation reactions. The cycloaddition precursors are flowed through these capillaries while the metal film is being heated to high temperatures using microwave irradiation. The transformation can be optimized rapidly, tolerates a wide number of functional groups, is highly regioselective, and proceeds in good to excellent conversion.
Sigma-Aldrich
1-Ethynyl-3-fluorobenzene, 98%