- Determination of benzotriazoles and benzothiazoles in human urine by liquid chromatography-tandem mass spectrometry.
Determination of benzotriazoles and benzothiazoles in human urine by liquid chromatography-tandem mass spectrometry.
Benzotriazole (BTR) and benzothiazole (BTH) derivatives are used in a wide variety of industrial and consumer products and have been reported to occur in the environment. Owing to a lack of analytical methods, human exposure to BTR and BTH is still unknown. In this study, a liquid chromatography-electrospray ionization tandem mass spectrometry (LC-ESI(+)MS/MS) method was developed for simultaneous determination of five 1,2,3-benzotriazoles and five 1,3-benzothiazoles in human urine. The target benzotriazoles were 1H-benzotriazole, 1-hydroxy-benzotriazole, tolyltriazole, xylyltriazole (or 5,6-dimethyl-1H-benzotriazole), and 5-chloro-benzotriazole, and the target benzothiazoles were benzothiazole, 2-hydroxy-benzothiazole, 2-methylthio-benzothiazole, 2-amino-benzothiazole, and 2-thiocyanomethylthio-benzothiazole. Urine specimens were enzymatically deconjugated with β-glucuronidase and extracted by a solid-phase extraction (SPE) procedure for the measurement of total concentrations (i.e., free + conjugated forms) of BTRs and BTHs. Additionally, a liquid-liquid extraction (LLE) method was developed for comparison of extraction efficiencies between SPE and LLE. The limits of detection (LODs) ranged from 0.07 (2-amino-benzothiazole) to 4.0 ng/mL (benzothiazole) for the SPE method and from 0.04 (tolyltriazole) to 6.4 ng/mL (benzothiazole) for the LLE method. A total of 100 urine specimens, collected from Athens, Greece, were analyzed by enzymatic deconjugation and SPE. Benzothiazole and tolyltriazole were found frequently, and their concentrations were on the order of a few ng/mL. To our knowledge, this is the first study on the occurrence of 10 BTR and BTH compounds in human urine.