Merck
  • Whole-mount fluorescent in situ hybridization staining of the colonial tunicate Botryllus schlosseri.
Whole-mount fluorescent in situ hybridization staining of the colonial tunicate Botryllus schlosseri.
Genesis (New York, N.Y. : 2000) (2014-09-03)
Adam D Langenbacher, Delany Rodriguez, Alessandro Di Maio, Anthony W De Tomaso
ABSTRACT
Botryllus schlosseri is a colonial ascidian with characteristics that make it an attractive model for studying immunology, stem cell biology, evolutionary biology, and regeneration. Transcriptome sequencing and the recent completion of a draft genome sequence for B. schlosseri have revealed a large number of genes, both with and without vertebrate homologs, but analyzing the spatial and temporal expression of these genes in situ has remained a challenge. Here, we report a robust protocol for in situ hybridization that enables the simultaneous detection of multiple transcripts in whole adult B. schlosseri using Tyramide Signal Amplification in conjunction with digoxigenin- and dinitrophenol-labeled RNA probes. Using this protocol, we have identified a number of genes that can serve as markers for developing and mature structures in B. schlosseri, permitting analysis of phenotypes induced in loss-of-function experiments.
Product Description
Roche
DIG RNA Labeling Mix, sufficient for 20 reactions, solution
Roche
Anti-Digoxigenin-POD, Fab fragments, from sheep
Roche
T7 RNA Polymerase, from Escherichia coli BL 21/pAR 1219
Roche
SP6 RNA Polymerase, from Escherichia coli BL 21/pSR3