Merck
  • Taste-active compound levels in Korean native chicken meat: The effects of bird age and the cooking process.
Taste-active compound levels in Korean native chicken meat: The effects of bird age and the cooking process.
Poultry science (2015-06-08)
Dinesh D Jayasena, Samooel Jung, Hyun Joo Kim, Hae In Yong, Ki Chang Nam, Cheorun Jo
ABSTRACT
The effects of bird age and the cooking process on the levels of several taste-active compounds, including inosine 5'-monophosphate (IMP), glutamic acid, cysteine, reducing sugars, as well as oleic, linoleic, arachidonic, and docosahexaenoic acids (DHA), in the breast and leg meats from a certified meat-type commercial Korean native chicken (KNC) strain (Woorimatdag) were investigated. KNC cocks were raised under similar standard conditions at a commercial chicken farm, and breast and leg meats from birds of various ages (10, 11, 12, 13, and 14 wk; 10 birds/age group) were obtained. After raw and cooked meat samples were prepared, they were analyzed for the aforementioned taste-active compounds. Compared to the leg meat, KNC breast meat had higher levels of IMP, arachidonic acid, and DHA, but lower levels of the other taste-active compounds (P < 0.05). KNC meat lost significant amounts of all the taste-active compounds, excluding oleic and linoleic acids, during the cooking process (P < 0.05). However, bird age only had a minor effect on the levels of these taste-active compounds. The results of this study provide useful information regarding the levels of taste-active compounds in KNC meat from birds of different ages, and their fate during the cooking process. This information could be useful for selection and breeding programs, and for popularizing native chicken meat.
Sigma-Aldrich
Trichloroacetic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%
Sigma-Aldrich
Trichloroacetic acid, ≥99.0% (titration)
Sigma-Aldrich
Trichloroacetic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Boron trifluoride, electronic grade, ≥99.99%
Sigma-Aldrich
Trichloroacetic acid, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Sucrose, SAJ first grade
Sigma-Aldrich
Sucrose, JIS special grade
Sigma-Aldrich
Trichloroacetic acid solution, 6.1 N
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, Grade I, ≥99% (GC), suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture