Merck
  • Self-Extinguishing Lithium Ion Batteries Based on Internally Embedded Fire-Extinguishing Microcapsules with Temperature-Responsiveness.
Self-Extinguishing Lithium Ion Batteries Based on Internally Embedded Fire-Extinguishing Microcapsules with Temperature-Responsiveness.
Nano letters (2015-07-16)
Taeeun Yim, Min-Sik Park, Sang-Gil Woo, Hyuk-Kwon Kwon, Jung-Keun Yoo, Yeon Sik Jung, Ki Jae Kim, Ji-Sang Yu, Young-Jun Kim
ABSTRACT
User safety is one of the most critical issues for the successful implementation of lithium ion batteries (LIBs) in electric vehicles and their further expansion in large-scale energy storage systems. Herein, we propose a novel approach to realize self-extinguishing capability of LIBs for effective safety improvement by integrating temperature-responsive microcapsules containing a fire-extinguishing agent. The microcapsules are designed to release an extinguisher agent upon increased internal temperature of an LIB, resulting in rapid heat absorption through an in situ endothermic reaction and suppression of further temperature rise and undesirable thermal runaway. In a standard nail penetration test, the temperature rise is reduced by 74% without compromising electrochemical performances. It is anticipated that on the strengths of excellent scalability, simplicity, and cost-effectiveness, this novel strategy can be extensively applied to various high energy-density devices to ensure human safety.
Sigma-Aldrich
Methyl methacrylate, 99%, stabilized
Sigma-Aldrich
Methyl methacrylate, SAJ first grade, ≥99.0%
Sigma-Aldrich
Methyl methacrylate, contains ≤30 ppm MEHQ as inhibitor, 99%
Sigma-Aldrich
Ethylene glycol dimethacrylate, 98%, contains 90-110 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Ethyl methyl carbonate, 98%
Sigma-Aldrich
Ethyl methyl carbonate, 99%