Merck
Five new bioactive compounds from Chenopodium ambrosioides.
Journal of Asian natural products research (2015-05-23)
Kun Song, Jian Zhang, Peng Zhang, Hong-Qing Wang, Chao Liu, Bao-Ming Li, Jie Kang, Ruo-Yun Chen
ABSTRACT
Five new bioactive compounds, chenopodiumamines A-D (1-4) and chenopodiumoside A (5), were isolated from the ethanol extract of Chenopodium ambrosioides. The structures of these compounds were elucidated by various spectroscopic means (UV, IR, HR-ESI-MS, 1D and 2D NMR). Compounds 1-3 had moderate antioxidant and anti-inflammatory activities.
Sigma-Aldrich
Water, BioPerformance Certified
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sigma-Aldrich
L-Cysteine methyl ester hydrochloride, 98%