Merck
  • Proteomic analysis of Pteropus alecto kidney cells in response to the viral mimic, Poly I:C.
Proteomic analysis of Pteropus alecto kidney cells in response to the viral mimic, Poly I:C.
Proteome science (2015-11-05)
Lawrence Mok, James W Wynne, Kris Ford, Brian Shiell, Antony Bacic, Wojtek P Michalski
ABSTRACT
Bats are recognised as an important reservoir for a number of highly pathogenic zoonotic viruses. While many of these viruses cause severe and often fatal disease in humans, bats are able to coexist with these viruses without clinical signs of disease. The mechanism conferring this antiviral response is not fully understood. Here, we investigated the differential protein expression of immortalised Pteropus alecto kidney cells (PaKiT03) following transfection with the viral mimic, Poly I:C. Two complementary proteomic approaches, difference gel electrophoresis (DIGE) and isobaric tagging for relative and absolute quantitation (iTRAQ) were used to quantify changes in protein expression following Poly I:C stimulation at 4, 8 and 20 hr post treatment (hpt). The expression of ISG54 gene, a known responder to virus infection and Poly I:C treatment, was significantly induced in transfected cells compared with mock-transfected cells. Through iTRAQ analysis we show that Poly I:C up-regulates key glycolytic enzymes at 4 hpt within PaKiT03 cells. In contrast, at 20 hpt PaKiT03 cells down-regulated ribosomal subunit proteins. The analysis with DIGE of Poly I:C transfected PaKiT03 cells showed over 215 individual spots differentially regulated, however only 25 spots could be unambiguously identified by LC-MS/MS. Immunoblotting confirmed the up-regulation of Eno1 and Tpi1 in PaKiT03 cells following Poly I:C transfection. A comparison with human cells (HEK293T and HeLa) and one additional bat cell line (PaLuT02), demonstrated that glycolytic pathways are also induced in these cell types, but at different intensities. The two techniques, DIGE and iTRAQ identified largely overlapping sets of differentially expressed proteins, however DIGE unambiguously identified significantly less proteins than iTRAQ. Poly I:C induced a rapid metabolic shift towards glycolysis within the PaKiT03 cells at 4 hpt, presumably as a consequence of increased energy requirements. On the other hand ribosomal subunit proteins were seen as down-regulated by iTRAQ, these proteins may be the limiting factors in the translational machinery available for virus replication. This study provides new insight into the antiviral response of bat cells, highlighting the importance of energy metabolism.
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Urea, SAJ first grade, ≥98.0%
Sigma-Aldrich
Formic acid, JIS special grade, ≥98.0%
Sigma-Aldrich
Urea, JIS special grade, ≥99.0%
Sigma-Aldrich
Urea, ≥99.0%
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
SAFC
L-Glutamine
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, BioUltra, for molecular biology, 99% (T)
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Pyridoxine, ≥98%
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium/Nutrient Mixture F-12 Ham, With L-glutamine, 15 mM HEPES, and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%