Merck
  • Down-regulation of osmotin (PR5) gene by virus-induced gene silencing (VIGS) leads to susceptibility of resistant Piper colubrinum Link. to the oomycete pathogen Phytophthora capsici Leonian.
Down-regulation of osmotin (PR5) gene by virus-induced gene silencing (VIGS) leads to susceptibility of resistant Piper colubrinum Link. to the oomycete pathogen Phytophthora capsici Leonian.
Indian journal of experimental biology (2015-07-15)
K Anu, K K Jessymol, M Chidambareswaren, G S Gayathri, S Manjula
ABSTRACT
Piper colubrinum Link., a distant relative of Piper nigrum L., is immune to the oomycete pathogen Phytophthora capsici Leonian that causes 'quick wilt' in cultivated black pepper (P. nigrum). The osmotin, PR5 gene homologue, earlier identified from P. colubrinum, showed significant overexpression in response to pathogen and defense signalling molecules. The present study focuses on the functional validation of P. colubrinum osmotin (PcOSM) by virus induced gene silencing (VIGS) using Tobacco Rattle Virus (TRV)-based vector. P. colubrinum plants maintained under controlled growth conditions in a growth chamber were infiltrated with Agrobacterium carrying TRV empty vector (control) and TRV vector carrying PcOSM. Three weeks post infiltration, viral movement was confirmed in newly emerged leaves of infiltrated plants by RT-PCR using TRV RNA1 and TRV RNA2 primers. Semi-quantitative RT-PCR confirmed significant down-regulation of PcOSM gene in TRV-PcOSM infiltrated plant compared with the control plants. The control and silenced plants were challenged with Phytophthora capsici which demonstrated that knock-down of PcOSM in P. colubrinum leads to increased fungal mycelial growth in silenced plants compared to control plants, which was accompanied by decreased accumulation of H2O2 as indicated by 3,3'-diaminobenzidine (DAB) staining. Thus, in this study, we demonstrated that Piper colubrinum osmotin gene is required for resisting P. capsici infection and has possible role in hypersensitive cell death response and oxidative burst signaling during infection.
Sigma-Aldrich
Ethanol, ≥99.5%
Sigma-Aldrich
Ethanol, JIS 300, ≥99.5%, for residue analysis
Sigma-Aldrich
Ethanol, ≥99.5%, SAJ super special grade
Sigma-Aldrich
Ethanol, JIS 1000, ≥99.5%, for residue analysis
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for absorption spectrum analysis
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for fluorescence
Sigma-Aldrich
MES solution, BioUltra, for molecular biology, 0.5 M in H2O
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
3′,5′-Dimethoxy-4′-hydroxyacetophenone, 97%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanesulfonic acid, 95%
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
MES, low moisture content, ≥99% (titration)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for HPLC
Sigma-Aldrich
Ethanol, 94.8-95.8%
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
Sigma-Aldrich
L-Thyroxine sodium salt pentahydrate, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Thyroxine sodium salt pentahydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)