Merck
  • Synthesis of Tertiary and Quaternary Amine Derivatives from Wood Resin as Chiral NMR Solvating Agents.
Synthesis of Tertiary and Quaternary Amine Derivatives from Wood Resin as Chiral NMR Solvating Agents.
Molecules (Basel, Switzerland) (2015-11-28)
Tiina Laaksonen, Sami Heikkinen, Kristiina Wähälä
ABSTRACT
Chiral tertiary and quaternary amine solvating agents for NMR spectroscopy were synthesized from the wood resin derivative (+)-dehydroabietylamine (2). The resolution of enantiomers of model compounds [Mosher's acid (3) and its n-Bu₄N salt (4)] (guests) by (+)-dehydroabietyl-N,N-dimethylmethanamine (5) and its ten different ammonium salts (hosts) was studied. The best results with 3 were obtained using 5 while with 4 the best enantiomeric resolution was obtained using (+)-dehydroabietyl-N,N-dimethylmethanaminium bis(trifluoromethane-sulfonimide) (6). The compounds 5 and 6 showed a 1:1 complexation behaviour between the host and guest. The capability of 5 and 6 to recognize the enantiomers of various α-substituted carboxylic acids and their n-Bu₄N salts in enantiomeric excess (ee) determinations was demonstrated. A modification of the RES-TOCSY NMR pulse sequence is described, allowing the enhancement of enantiomeric discrimination when the resolution of multiplets is insufficient.
Sigma-Aldrich
Mercury, SAJ first grade, ≥99.5%
Sigma-Aldrich
Mercury, JIS special grade, ≥99.5%
Sigma-Aldrich
(+)-Dehydroabietylamine, technical grade, 60%
Sigma-Aldrich
Mercury, ACS reagent, 99.9995% trace metals basis