Merck
  • Cynara scolymus affects malignant pleural mesothelioma by promoting apoptosis and restraining invasion.
Cynara scolymus affects malignant pleural mesothelioma by promoting apoptosis and restraining invasion.
Oncotarget (2015-07-03)
Claudio Pulito, Federica Mori, Andrea Sacconi, Luca Casadei, Maria Ferraiuolo, Maria Cristina Valerio, Raffaela Santoro, Frauke Goeman, Anna Maidecchi, Luisa Mattoli, Cesare Manetti, Silvia Di Agostino, Paola Muti, Giovanni Blandino, Sabrina Strano
ABSTRACT
Malignant pleural mesothelioma is a poorly treated neoplasia arising from the pleural mesothelial lining. Here we document that the leaf extract of Cynara scolymus exerts broad antitumoral effects both in vitro and in vivo on mesothelioma cell lines. We found that Cynara scolymus treatment affects strongly cell growth, migration and tumor engraftment of mesothelioma cell lines. Strikingly, dietary feeding with Cynara scolymus leaf extract reduces the growth of mesothelioma xenografted tumors similarly to pemetrexed, a commonly employed drug in the treatment of mesothelioma. In aggregate our findings suggest that leaf extract of Cynara scolymus holds therapeutic potential for the treatment of mesothelioma.
Sigma-Aldrich
Sodium phosphate tribasic dodecahydrate, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Sodium phosphate tribasic dodecahydrate, ≥98%
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Sigma-Aldrich
trans-Platinum(II)diammine dichloride
Sigma-Aldrich
DAPI, for nucleic acid staining