Merck
  • Wet-chemical synthesis and applications of non-layer structured two-dimensional nanomaterials.
Wet-chemical synthesis and applications of non-layer structured two-dimensional nanomaterials.
Nature communications (2015-08-26)
Chaoliang Tan, Hua Zhang
ABSTRACT
Non-layer structured nanomaterials with single- or few-layer thickness have two-dimensional sheet-like structures and possess intriguing properties. Recent years have seen major advances in development of a host of non-layer structured ultrathin two-dimensional nanomaterials such as noble metals, metal oxides and metal chalcogenides. The wet-chemical synthesis has emerged as the most promising route towards high-yield and mass production of such nanomaterials. These nanomaterials are now finding increasing applications in a wide range of areas including catalysis, energy production and storage, sensor and nanotherapy, to name but a few.
Sigma-Aldrich
Diethylenetriamine, ReagentPlus®, 99%
Sigma-Aldrich
Diethylenetriamine, SAJ first grade, ≥98.0%
Sigma-Aldrich
Oleylamine, technical grade, 70%
SAFC
Oleylamine, ≥98% (primary amine)