Merck
  • Interaction of polyamines and mTOR signaling in the synthesis of antizyme (AZ).
Interaction of polyamines and mTOR signaling in the synthesis of antizyme (AZ).
Cellular signalling (2015-06-21)
Ramesh M Ray, Mitul Bavaria, Leonard R Johnson
ABSTRACT
Tissue polyamine levels are largely determined by the activity of ornithine decarboxylase (ODC, EC 4.1.17), which catalyzes the conversion of ornithine to the diamine putrescine. The activity of the enzyme is primarily regulated by a negative feedback mechanism involving ODC antizyme (AZ). Our previous studies demonstrated that AZ synthesis is stimulated by the absence of amino acids, the levels of which are sensed by the mTOR complex containing TORC1, which is stimulated by amino acids and inhibited by their absence, and TORC2 the function of which is not well defined. Polyamines, which cause a +1 ribosomal frameshift during the translation of AZ mRNA are required to increase AZ synthesis in both the presence and absence of amino acids. Amino acid starvation increases TORC2 activity. We have demonstrated that mTORC2 activity is necessary for AZ synthesis in the absence of amino acids. Tuberous sclerosis protein (TSC), a negative regulator of mTOR function regulates the activities of both the TORC1 and TORC2. TSC2 knockdown increased mTORC1 activity with concomitant inhibition of mTORC2 activity eliminating AZ induction in the absence of amino acids as well as that induced by spermidine. Thus, these results clearly demonstrate that in addition to polyamines, mTORC2 activity is necessary for AZ synthesis. Moreover, our results support a role for mTORC2 in the synthesis of a specific protein, AZ, which regulates growth of intestinal epithelial cells.
Sigma-Aldrich
Spermine, ≥99.0% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Spermidine, BioUltra, for molecular biology, ≥99.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Spermidine, ≥99% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Spermine, ≥97%
Pricing and availability is not currently available.
Sigma-Aldrich
L-Asparagine, BioReagent, suitable for cell culture, suitable for insect cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Spermidine, BioReagent, for molecular biology, suitable for cell culture, ≥98%
Pricing and availability is not currently available.
Sigma-Aldrich
Spermine, BioReagent, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
L-Asparagine, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
1,4-Diaminobutane, 99%
Pricing and availability is not currently available.