Merck
CARMA3 Is Critical for the Initiation of Allergic Airway Inflammation.
Journal of immunology (Baltimore, Md. : 1950) (2015-06-05)
Benjamin Causton, Ravisankar A Ramadas, Josalyn L Cho, Khristianna Jones, Ana Pardo-Saganta, Jayaraj Rajagopal, Ramnik J Xavier, Benjamin D Medoff
ABSTRACT
Innate immune responses to allergens by airway epithelial cells (AECs) help initiate and propagate the adaptive immune response associated with allergic airway inflammation in asthma. Activation of the transcription factor NF-κB in AECs by allergens or secondary mediators via G protein-coupled receptors (GPCRs) is an important component of this multifaceted inflammatory cascade. Members of the caspase recruitment domain family of proteins display tissue-specific expression and help mediate NF-κB activity in response to numerous stimuli. We have previously shown that caspase recruitment domain-containing membrane-associated guanylate kinase protein (CARMA)3 is specifically expressed in AECs and mediates NF-κB activation in these cells in response to stimulation with the GPCR agonist lysophosphatidic acid. In this study, we demonstrate that reduced levels of CARMA3 in normal human bronchial epithelial cells decreases the production of proasthmatic mediators in response to a panel of asthma-relevant GPCR ligands such as lysophosphatidic acid, adenosine triphosphate, and allergens that activate GPCRs such as Alternaria alternata and house dust mite. We then show that genetically modified mice with CARMA3-deficient AECs have reduced airway eosinophilia and proinflammatory cytokine production in a murine model of allergic airway inflammation. Additionally, we demonstrate that these mice have impaired dendritic cell maturation in the lung and that dendritic cells from mice with CARMA3-deficient AECs have impaired Ag processing. In conclusion, we show that AEC CARMA3 helps mediate allergic airway inflammation, and that CARMA3 is a critical signaling molecule bridging the innate and adaptive immune responses in the lung.
Sigma-Aldrich
2,3-Dimercapto-1-propanol, ≥98% (iodometric)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Red, DUOLINK® : PLA kit for Flow Cytometry with Red Detection
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Green, Duolink® PLA kit for Flow Cytometry with Green Detection
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Mouse/Goat
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® In Situ Microplate Heat Transfer Block
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® In Situ Detection Reagents Red
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® In Situ Probemaker PLUS
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® In Situ Microplate Nuclear Stain, Anti-Fade
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse MINUS, Affinity purified Donkey anti-Mouse IgG (H+L)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® In Situ Mounting Medium with DAPI
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Goat MINUS, Affinity purified Donkey anti-Goat IgG (H+L)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit PLUS, Affinity purified Donkey anti-Rabbit IgG (H+L)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Goat/Rabbit
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Goat/Rabbit
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Mouse/Goat
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Mouse/Rabbit
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
MISSION® esiRNA, targeting human CARD10
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® In Situ Detection Reagents Orange
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® In Situ Probemaker MINUS
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse PLUS
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® In Situ Detection Reagents Brightfield
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® In Situ Detection Reagents FarRed
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Goat PLUS
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit MINUS
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® In Situ Detection Reagents Green
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Aluminum potassium sulfate dodecahydrate, BioXtra, ≥98.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Aluminum potassium sulfate dodecahydrate, BioReagent, suitable for insect cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® In Situ Wash Buffers, Fluorescence
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® In Situ Wash Buffer, Brightfield
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® PLA Control Kit - PPI, Control Kit to reliably detect Protein-Protein Interactions (PPI) with Duolink® PLA