Merck
  • Angiotensin-converting enzyme gene insertion deletion (ACE I/D) polymorphism in Saudi children with congenital heart disease.
Angiotensin-converting enzyme gene insertion deletion (ACE I/D) polymorphism in Saudi children with congenital heart disease.
European review for medical and pharmacological sciences (2015-07-01)
N M Alazhary, M M F Morsy, K M Al-Harbi
ABSTRACT
Congenital heart diseases (CHDs) are the leading cause of infant deaths worldwide. Angiotensin-converting enzyme (ACE) gene I/D polymorphism is associated with many cardiovascular diseases. The precise relationship between this polymorphism and CHDs is not clear. The aim of this work is to determine the normal distribution of I/D polymorphism in Saudi citizens and to test for any association between this polymorphism and CHDs in Saudi children. Ninety-six CHD cases and 145 controls were included in this study. DNA was isolated from their peripheral blood, and then ACE I/D gene polymorphism was assayed by polymerase chain reaction (PCR). There was no significant difference among the frequencies of the DD, DI and II genotypes in patients and controls [39 (41%), 64 (44%), 48 (51%) and 62 (43%), 7 (7%), 19 (13%)] respectively (p-value = 0.3 and OR (95% CI) = 0.3). There was no significant difference between D allele (DD+DI) and II genotype distribution among patients and controls [p-value = 0.2 & OR (95% CI) = 1.9 (0.8-4.7)]. Moreover, there was no difference between I allele (II+DI) and DD frequency [p-value = 0.8 & OR = 0.9, CI = 0.5-1.5]. ACE I/D gene polymorphism is not associated with CHDs in Saudi children. Further large-scale studies are necessary to establish our findings.
Sigma-Aldrich
Ethidium bromide, ≥95.0%
Sigma-Aldrich
Ethidium bromide, BioReagent, for molecular biology, powder
Sigma-Aldrich
Ethidium bromide, ~95% (HPLC)
Sigma-Aldrich
Ethidium bromide solution, 10 mg/mL
Supelco
Pyridaben, PESTANAL®, analytical standard
Sigma-Aldrich
Ethidium bromide solution, for fluorescence, ~1% in H2O
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 500 μg/mL in H2O
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 10 mg/mL in H2O