- Novel EXOSC3 mutation causes complicated hereditary spastic paraplegia.
Novel EXOSC3 mutation causes complicated hereditary spastic paraplegia.
We describe two pairs of siblings from a consanguineous family manifesting autosomal recessive hereditary spastic paraplegia caused by a novel mutation in the EXOSC3 gene, previously reported in pontocerebellar hypoplasia type 1. Clinical findings included delayed motor milestones, early-onset spastic paraplegia, variable cognitive disability, and cerebellar signs. Cerebral imaging demonstrated enlarged cisterna magna and mild hypoplasia and atrophy of the lower vermis with a normal pons. Genetic analysis using homozygosity mapping followed by whole exome sequencing identified homozygous c.571G>T; p.G191C mutation in the EXOSC3 gene. We suggest that EXOSC3 mutations may present not only as pontocerebellar hypoplasia type 1, but also as a complicated form of hereditary spastic paraplegia without pontine hypoplasia or atrophy.