Merck
  • Germline and somatic mutations in cyclin-dependent kinase inhibitor genes CDKN1A, CDKN2B, and CDKN2C in sporadic parathyroid adenomas.
Germline and somatic mutations in cyclin-dependent kinase inhibitor genes CDKN1A, CDKN2B, and CDKN2C in sporadic parathyroid adenomas.
Hormones & cancer (2013-05-30)
Jessica Costa-Guda, Chen-Pang Soong, Vaishali I Parekh, Sunita K Agarwal, Andrew Arnold
ABSTRACT
The molecular pathogenesis of sporadic parathyroid adenomas is incompletely understood. The possible role of cyclin-dependent kinase inhibitor (CDKI) genes was raised by recognition of cyclin D1 as a parathyroid oncogene, identification of rare germline mutations in CDKI genes in patients with multiple endocrine neoplasia type 1; that in rodents, mutation in Cdkn1b caused parathyroid tumors; and subsequently through identification of rare predisposing germline sequence variants and somatic mutation of CDKN1B, encoding p27(kip1), in sporadic human parathyroid adenoma. We therefore sought to determine whether mutations/variants in the other six CDKI genes CDKN1A, CDKN1C, CDKN2A, CDKN2B, CDKN2C, and CDKN2D, encoding p21, p57, p14(ARF)/p16, p15, p18, and p19, respectively, contribute to the development of typical parathyroid adenomas. In a series of 85 sporadic parathyroid adenomas, direct DNA sequencing identified alterations in five adenomas (6 %): Two contained distinct heterozygous changes in CDKN1A, one germline and one of undetermined germline status; one had a CDKN2B germline alteration, accompanied by loss of the normal allele in the tumor (LOH); two had variants of CDKN2C, one somatic and one germline with LOH. Abnormalities of three of the mutant proteins were readily demonstrable in vitro. Thus, germline mutations/rare variants in CDKN1A, CDKN2B, and CDKN2C likely contribute to the development of a significant subgroup of common sporadic parathyroid adenomas, and somatic mutation in CDKN2C further suggests a direct role for CDKI alteration in conferring a selective growth advantage to parathyroid cells, providing novel support for the concept that multiple CDKIs can play primary roles in human neoplasia.
Sigma-Aldrich
Anti-Neomycin Phosphotransferase II Antibody, Upstate®, from rabbit
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-α-Tubulin Mouse mAb (DM1A), liquid, clone DM1A, Calbiochem®