Merck
  • A neurally enriched coronin-like protein, ClipinC, is a novel candidate for an actin cytoskeleton-cortical membrane-linking protein.
A neurally enriched coronin-like protein, ClipinC, is a novel candidate for an actin cytoskeleton-cortical membrane-linking protein.
The Journal of biological chemistry (1999-05-01)
T Nakamura, K Takeuchi, S Muraoka, H Takezoe, N Takahashi, N Mori
ABSTRACT
Brain-enriched human FC96 protein shows a close sequence similarity to the Dictyostelium actin-binding protein coronin, which has been implicated in cell motility, cytokinesis, and phagocytosis. A phylogenetic tree analysis revealed that FC96 and two other mammalian molecules (p57 and IR10) form a new protein family, the coronin-like protein (Clipin) family; thus hereafter we refer to FC96 as ClipinC. A WD domain and a succeeding alpha-helical region are conserved among coronin and Clipin family members. ClipinC is predominantly expressed in the brain, and discrete areas in the mouse brain were intensely labeled with anti-ClipinC antibodies. ClipinC was also shown to bind directly to F-actin in vitro. Immunocytochemical analysis revealed that ClipinC accumulated at focal adhesions as well as at neurite tips and stress fibers. Furthermore, ClipinC was associated with vinculin, which is a major component of focal contacts. These results indicate that ClipinC is also a component part of the cross-bridge between the actin cytoskeleton and the plasma membrane. These findings and the previously reported function of coronin suggest that ClipinC may play specific roles in the reorganization of neuronal actin structure, a change that has been implicated in both cell motility and growth cone advance.