- TECPR2: a new autophagy link for neurodegeneration.
TECPR2: a new autophagy link for neurodegeneration.
Autophagy dysfunction has been implicated in a group of progressive neurodegenerative diseases, and has been reported to play a major role in the pathogenesis of these disorders. We have recently reported a recessive mutation in TECPR2, an autophagy-implicated WD repeat-containing protein, in five individuals with a novel form of monogenic hereditary spastic paraparesis (HSP). We found that diseased skin fibroblasts had a decreased accumulation of the autophagy-initiation protein MAP1LC3B/LC3B, and an attenuated delivery of both LC3B and the cargo-recruiting protein SQSTM1/p62 to the lysosome where they are subject to degradation. The discovered TECPR2 mutation reveals for the first time a role for aberrant autophagy in a major class of Mendelian neurodegenerative diseases, and suggests mechanisms by which impaired autophagy may impinge on a broader scope of neurodegeneration.