Merck
  • Cord blood serum-based eye drops: the impact of donor haematological and obstetric factors on the variability of epidermal growth factor levels.
Cord blood serum-based eye drops: the impact of donor haematological and obstetric factors on the variability of epidermal growth factor levels.
Blood transfusion = Trasfusione del sangue (2013-10-15)
Piera Versura, Marina Buzzi, Giuseppe Giannaccare, Marco Grillini, Adriana Terzi, Pasqualepaolo Pagliaro, Emilio C Campos
ABSTRACT
Cord blood serum (CBS)-based eye drops are successfully used in corneal epithelial wound healing and are prepared to supply a known amount of epidermal growth factor (EGF). Product standardisation includes expensive EGF dosage in all cord blood (CB) units. The influence of donor obstetric and haematological characteristics on EGF content was evaluated, to exclude unsuitable CBS and pre-select those CB units able to provide the correct EGF supply for healing corneal wounds. Data were retrospectively collected from 135 donors included in the Emilia Romagna Cord Blood Bank records. Obstetric characteristics, parity and gestational age of the mother, sex, birth weight and Apgar score of the neonate, placental weight, duration of labour and mode of delivery were considered. Haematological characteristics, CD34+ cell number, and total nucleated cell, white blood cell and platelet counts were recorded. EGF content in CB units was estimated by enzyme-linked immunosorbent assay. Statistical evaluation was performed by Mann-Whitney unpaired and Student's t tests. Correlations between variables were evaluated by using Pearson's (r) or Spearman's (ρ) correlation coefficients. EGF content was significantly higher in CBS from donors aged <30 years and after vaginal deliveries as compared with scheduled Caesarean sections (1,386±580 vs 1,106±391 pg/mL; P=0.002). EGF content was significantly correlated with duration of labour (r=0.45; P=0.0001), number of CD34+ cells/mL (r=0.3; P=0.002) particularly in vaginal deliveries (r=0.36; P=0.003), mother's age (-0.25; P=0.005), neonate's birth weight (r=0.27; P=0.005), and total nucleated cell (r=0.25; P=0.006), white cell (r=0.29; P=0.001) and platelet (r=0.24; P=0.009) counts. No significant correlations were found between EGF content and parity, gestational age, placental weight, neonate's sex or Apgar scores. EGF levels are higher in CB units from younger mothers (<30 years), with longer labour duration (>6 hours), and higher CD34+ cell content (>0.05×10(6)/mL). In order to optimise the preparation and costs of CBS-based eye drops, pre-selection of CB units is recommended.
Product Description
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
USP
Dextrose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)