Merck
  • MicroRNA-590 attenuates lipid accumulation and pro-inflammatory cytokine secretion by targeting lipoprotein lipase gene in human THP-1 macrophages.
MicroRNA-590 attenuates lipid accumulation and pro-inflammatory cytokine secretion by targeting lipoprotein lipase gene in human THP-1 macrophages.
Biochimie (2014-08-26)
Ping-Ping He, Xin-Ping Ouyang, Yan-Yan Tang, Li Liao, Zong-Bao Wang, Yun-Cheng Lv, Guo-Ping Tian, Guo-Jun Zhao, Liang Huang, Feng Yao, Wei Xie, Yu Lin Tang, Wu-Jun Chen, Min Zhang, Yuan Li, Jian-Feng Wu, Juan Peng, Xiang-Yu Liu, Xi-Long Zheng, Wei-Dong Yin, Chao-Ke Tang
ABSTRACT
Accumulating evidence suggests that microRNA-590 (miR-590) has protective effects on cardiovascular diseases, but the mechanism is unknown. Interestingly, previous studies from our laboratory and others have shown that macrophage-derived lipoprotein lipase (LPL) might accelerate atherosclerosis by promoting lipid accumulation and inflammatory response. However, the regulation of LPL at the post-transcriptional level by microRNAs has not been fully understood. In this study, we explored whether miR-590 affects the expression of LPL and its potential subsequent effects on lipid accumulation and pro-inflammatory cytokine secretion in human THP-1 macrophages. Using bioinformatics analyses and dual-luciferase reporter assays, we found that miR-590 directly inhibited LPL protein and mRNA expression by targeting LPL 3'UTR. LPL Activity Assays showed that miR-590 reduced LPL activity in the culture media. Oil Red O staining and high-performance liquid chromatography assays showed that miR-590 had inhibitory effects on the lipid accumulation in human THP-1 macrophages. We also illustrated that miR-590 alleviated pro-inflammatory cytokine secretion in human THP-1 macrophages as measured by ELISA. With the method of small interfering RNA, we found that LPL siRNA can inhibit the miR-590 inhibitor-induced increase in lipid accumulation and secretion of pro-inflammatory cytokines in oxLDL-treated human THP-1 macrophages. MiR-590 attenuates lipid accumulation and pro-inflammatory cytokine secretion by targeting LPL gene in human THP-1 macrophages. Therefore, targeting miR-590 may offer a promising strategy to treat atherosclerotic cardiovascular diseases.
Sigma-Aldrich
MISSION® esiRNA, targeting human LPL
Sigma-Aldrich
MISSION® esiRNA, targeting human LCP1
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Lpl
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Lcp1