Merck
  • Cytoplasmic maspin expression predicts poor prognosis of patients with soft tissue sarcomas.
Cytoplasmic maspin expression predicts poor prognosis of patients with soft tissue sarcomas.
Diagnostic pathology (2014-11-02)
Chikako Takeda, Yuzo Takagi, Tatsushi Shiomi, Kanae Nosaka, Hideki Yamashita, Mari Osaki, Koji Endo, Takeshi Minamizaki, Ryota Teshima, Hideki Nagashima, Yoshihisa Umekita
ABSTRACT
Maspin is a 42 kDa protein known to act as a tumor suppressor. Although its function has not been fully elucidated, numerous reports have investigated the prognostic impact of maspin in patients with several types of cancer. However, there have been no reports on the association between maspin expression and the prognosis of patients with soft tissue sarcomas (STS). The aim of this study was thus to explore the association of maspin expression with the prognosis of patients with STS. One-hundred and eight paraffin-embedded STS tissue samples were immunohistochemically analyzed using antibodies for maspin and Ki-67 antigen. The patients were followed up for 1 to 300 months (median: 33 months) and the prognostic value was evaluated by log-rank test and Cox's regression hazard model. Cytoplasmic maspin expression was observed in 48.1% of specimens, and was significantly correlated with a higher FNCLCC grade (P = 0.002) and the presence of distant metastases (P = 0.001), and those with cytoplasmic maspin expression had both shorter disease-free survival (DFS) and overall survival (OS) by log-rank test (P <0.001, P = 0.001, respectively). By Cox's multivariate analysis, the presence of distant metastases was the only prognostic factor for DFS and OS. This is the first report to reveal an association between maspin expression and the prognosis of patients with STS. Although further studies with a larger series of patients and a longer follow-up period will be needed, cytoplasmic maspin expression could be an indicator of unfavorable prognosis in patients with STS. The virtual slide(s) for this article can be found here: http://www.diagnosticpathology.diagnomx.eu/vs/13000_2014_205.
Sigma-Aldrich
Maspin human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
MISSION® esiRNA, targeting human SERPINB5