Merck
  • Vitamin D status modifies the association between statin use and musculoskeletal pain: a population based study.
Vitamin D status modifies the association between statin use and musculoskeletal pain: a population based study.
Atherosclerosis (2014-12-02)
Travis Y Morioka, Alice J Lee, Suzanne Bertisch, Catherine Buettner
ABSTRACT
Past studies examining the effect of vitamin D on statin myalgia have been variable; however, these studies were done in limited samples not representative of the general population. We aimed to evaluate whether vitamin D status modifies the association between statin use and musculoskeletal pain in a sample representative of the general population. We conducted a cross-sectional study using the National Health and Nutrition Examination Survey 2001-2004. Musculoskeletal symptoms and statin use were self-reported. Vitamin D status was assessed using serum 25 hydroxyvitamin D (25[OH]D), categorized as <15 ng/mL or ≥15 ng/mL. To evaluate if vitamin D status modifies the association between statin use and prevalent musculoskeletal pain, we performed multivariable-adjusted logistic regression models stratified by 25(OH)D status. Among 5907 participants ≥40 years old, mean serum 25(OH)D was 23.6 ng/mL (95% CI, 22.9-24.3). In stratified multivariable-adjusted logistic regression models, individuals with 25(OH)D <15 ng/mL, using a statin had a significantly higher odds of musculoskeletal pain compared to those not using a statin (adjusted odds ratio [aOR], 1.90; 95% CI, 1.18-3.05). Among those with 25(OH)D ≥15 ng/mL, we found no significant association between statin use and musculoskeletal pain (aOR, 0.91; 95% CI, 0.71-1.16). Among adults ≥ 40 years old with 25(OH)D <15 ng/mL, statin users had nearly 2 times greater odds of reporting musculoskeletal pain compared to non-statin users. Our findings support the hypothesis that vitamin D deficiency modifies the risk of musculoskeletal symptoms experienced with statin use.
Sigma-Aldrich
Bromocresol Purple, SAJ special grade
Sigma-Aldrich
Bromocresol Purple, BioReagent, suitable for indicator, Dye content 90 %
Sigma-Aldrich
Bromocresol Purple, Technical grade