Merck
  • Effect of selenite on T-cell mitogenesis: contribution of ROS production and apoptosis signal-regulating kinase 1.
Effect of selenite on T-cell mitogenesis: contribution of ROS production and apoptosis signal-regulating kinase 1.
Biological & pharmaceutical bulletin (2014-08-05)
Hitoshi Ueno, Hitomi Kajihara, Hajime Nakamura, Tomofumi Okuno, Fumitoshi Sakazaki, Tomohiro Arakawa, Hirofumi Ogino, Katsuhiko Nakamuro, Junji Yodoi
ABSTRACT
Although supplementation with the selenocompound, sodium selenite has been shown to stimulate the concanavalin A-induced T-cell mitogenic response, the mechanisms responsible remain unclear. This study was conducted to evaluate the relationships between the induction of apoptosis, formation of tumor necrosis factor (TNF)-alpha and reactive oxygen species (ROS), activation of apoptosis signal-regulating kinase (ASK) 1 and the thioredoxin (Trx) system when mitogenesis was stimulated by selenite. TNF-alpha was dose-dependently released by mouse splenocytes treated with selenite, and apoptosis was induced when TNF-alpha was added at the indicated concentrations. However, supplementation with selenite at low concentrations inhibited the accumulation of ROS with the increased expression of Trx reductase 1 and induction of apoptosis in wild-type splenocytes, and also at high concentrations in Trx-1-transgenic mouse splenocytes. The suppression of apoptosis was accompanied by a decrease in the expression of phospho-ASK1. These results suggest that the stimulation of T-cell mitogenesis by selenite may be partly attributed to the inhibited accumulation of ROS due to a reduced Trx-1/TR1 system, the inactivation of ASK1, and the suppression of apoptosis.
Sigma-Aldrich
Sodium selenite, SAJ first grade, ≥90.0%
Sigma-Aldrich
Sodium selenite, anhydrous, ≥90.0% (RT)
Sigma-Aldrich
Sodium selenite, γ-irradiated, lyophilized powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium selenite, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Sodium selenite, 99%