Merck
  • Differential expression of centrosome regulators in Her2+ breast cancer cells versus non-tumorigenic MCF10A cells.
Differential expression of centrosome regulators in Her2+ breast cancer cells versus non-tumorigenic MCF10A cells.
Cell division (2014-10-04)
Mi-Young Lee, Mihaela Marina, Jamie L King, Harold I Saavedra
ABSTRACT
Centrosome amplification (CA) amongst particular breast cancer subtypes (Her2+ subtype) is associated with genomic instability and aggressive tumor phenotypes. However, changes in signaling pathways associated with centrosome biology have not been fully explored in subtype specific models. Novel centrosome regulatory genes that are selectively altered in Her2+ breast cancer cells are of interest in discerning why CA is more prevalent in this subtype. To determine centrosome/cell cycle genes that are altered in Her2+ cells that display CA (HCC1954) versus non-tumorigenic cells (MCF10A), we carried out a gene microarray. Expression differences were validated by real-time PCR and Western blotting. After the microarray validation, we pursued a panel of upregulated and downregulated genes based on novelty/relevance to centrosome duplication. Functional experiments measuring CA and BrdU incorporation were completed after genetic manipulation of targets (TTK, SGOL1, MDM2 and SFRP1). Amongst genes that were downregulated in HCC1954 cells, knockdown of MDM2 and SFRP1 in MCF10A cells did not consistently induce CA or impaired BrdU incorporation. Conversely, amongst upregulated genes in HCC1954 cells, knockdown of SGOL1 and TTK decreased CA in breast cancer cells, while BrdU incorporation was only altered by SGOL1 knockdown. We also explored the Kaplan Meier Plot resource and noted that MDM2 and SFRP1 are positively associated with relapse free survival in all breast cancer subtypes, while TTK is negatively correlated with overall survival of Luminal A patients. Based on this functional screen, we conclude that SGOL1 and TTK are important modulators of centrosome function in a breast cancer specific model.
Sigma-Aldrich
MISSION® esiRNA, targeting human TTBK2
Pricing and availability is not currently available.
Sigma-Aldrich
MISSION® esiRNA, targeting human TTK
Pricing and availability is not currently available.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Ttk
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-BrdU (Ab-3) Mouse mAb (Mobu-1), liquid, clone Mobu-1, Calbiochem®
Pricing and availability is not currently available.