Merck
  • NEO212, temozolomide conjugated to perillyl alcohol, is a novel drug for effective treatment of a broad range of temozolomide-resistant gliomas.
NEO212, temozolomide conjugated to perillyl alcohol, is a novel drug for effective treatment of a broad range of temozolomide-resistant gliomas.
Molecular cancer therapeutics (2014-07-06)
Hee-Yeon Cho, Weijun Wang, Niyati Jhaveri, David Jungpa Lee, Natasha Sharma, Louis Dubeau, Axel H Schönthal, Florence M Hofman, Thomas C Chen
ABSTRACT
Patients with glioblastoma multiforme (GBM), a malignant primary brain tumor, inevitably develop resistance to standard-of-care chemotherapy, temozolomide. This study explores the effects of the novel agent NEO212, a conjugate of temozolomide to perillyl alcohol, on temozolomide-resistant gliomas. NEO212 was tested for cytotoxic activity on three human temozolomide-resistant glioma cell lines, which were resistant to temozolomide based on overexpression of the base excision repair (BER) pathway, mismatch repair (MMR) deficiency, or overexpression of O(6) methyl-guanine-DNA methyltransferase (MGMT). BER expression was evaluated by Western blotting and PARP activity. MMR deficiency was determined by Western blotting and microsatellite instability. MGMT overexpression was evaluated by Western blotting and O(6)-benzylguanine (O(6)BG) inhibition. For in vivo evaluation of NEO212, temozolomide-resistant glioma cells were implanted into immune-incompetent mice, and NEO212 was administered. NEO212, at equimolar concentrations of temozolomide, was more cytotoxic for temozolomide-resistant cells than temozolomide and not toxic to normal cells. NEO212-induced cell death in temozolomide-resistant glioma cells was independent of such mechanisms of resistance as high levels of MGMT, MMR deficiencies, or overexpression of BER proteins. NEO212 functions as a DNA alkylating agent, similar to temozolomide; however, this novel conjugate is unique for it may induce endoplasmic reticulum (ER) stress and inhibits autophagy. In vivo studies show that NEO212 reduces intracranial tumor growth and increases animal survival without significant toxicity. These results demonstrate that NEO212 is an effective drug against malignant gliomas that can be used for a broad range of newly diagnosed and temozolomide-resistant gliomas.
Product Description
Supelco
Temozolomide, VETRANAL®, analytical standard
Sigma-Aldrich
Temozolomide, ≥98% (HPLC)
Supelco
Temozolomide, Pharmaceutical Secondary Standard; Certified Reference Material